5-Propyl-1H-pyrazol-3-ylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Propyl-1H-pyrazol-3-amine

H50969-03 H50969-MD
AAH50969-03EA 281.99 USD
AAH50969-03 AAH50969-MD
5-Propyl-1H-pyrazol-3-ylamine 97%
5-Propyl-1H-pyrazol-3-ylamine
Formula: C₆H₁₁N₃
MW: 125.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08061095
CAS Number: 126748-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR