3-(Pentafluorothio)benzoic acid 97%

Danger

Synonyms: 3-Sulphurpentafluorobenzoic acid3-Sulfurpentafluorobenzoic acid3-Carboxyphenylsulphur pentafluoride3-Carboxyphenylsulfur pentafluoride

H34499-06 H34499-03
AAH34499-03EA 99.5 USD
AAH34499-03 AAH34499-06
3-(Pentafluorothio)benzoic acid 97%
3-(Pentafluorothio)benzoic acid
Formula: C₇H₅F₅O₂S
MW: 248.17 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Ambient
MDL Number: MFCD03788519
CAS Number: 783240-27-7
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR