2-Fluoro-4-(pentafluorothio)benzoyl chloride 97%

Danger

H34481-03 H34481-MD
AAH34481-03EA 114.33 USD
AAH34481-03 AAH34481-MD
2-Fluoro-4-(pentafluorothio)benzoyl chloride 97%
2-Fluoro-4-(pentafluorothio)benzoyl chloride
Formula: C₇H₃ClF₆OS
MW: 284.60
Storage Temperature: Ambient
MDL Number: MFCD16652505
CAS Number: 1240256-85-2
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR