3-Bromo-5-(pentafluorothio)aniline 97%

Warning

H34456-MD H34456-03
AAH34456-03EA 198.69 USD
AAH34456-03 AAH34456-MD
3-Bromo-5-(pentafluorothio)aniline 97%
3-Bromo-5-(pentafluorothio)aniline
Formula: C₆H₅BrF₅NS
MW: 298.07
Storage Temperature: Ambient
MDL Number: MFCD16652416
CAS Number: 1240257-95-7
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR