6-Bromoimidazo[1,2-a]pyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H34453-03 H34453-06
AAH34453-03EA 87.79 USD
AAH34453-03 AAH34453-06
6-Bromoimidazo[1,2-a]pyridine 98%
6-Bromoimidazo[1,2-a]pyridine
Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD06496200
CAS Number: 6188-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR