8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine 97%

Warning

H34451-03 H34451-06
AAH34451-06EA 144.3 USD
AAH34451-06 AAH34451-03
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine 97%
8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine
Formula: C₈H₄ClF₃N₂
Melting Pt: 100…103 °C
Storage Temperature: Ambient
MDL Number: MFCD06496224
CAS Number: 178488-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR