3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one 96%

Warning

H34437-06 H34437-MD H34437-03
AAH34437-03EA 90.66 USD
AAH34437-03 AAH34437-06 AAH34437-MD
3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one 96%
3-Allyl-5,6-dimethyl-2-thioxo-2,3-dihydrothieno[2,3-d]pyrimidin-4(1H)-one
Formula: C₁₁H₁₂N₂OS₂
MW: 252.36 g/mol
Melting Pt: 202…203 °C
Storage Temperature: Ambient
MDL Number: MFCD01911592
CAS Number: 51486-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR