4-Bromopyridine-2-carbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-2-cyanopyridine

H34424-03 H34424-MD
AAH34424-03EA 201.99 USD
AAH34424-03 AAH34424-MD
4-Bromopyridine-2-carbonitrile 97%
4-Bromopyridine-2-carbonitrile
Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 87…91 °C
Storage Temperature: Ambient
MDL Number: MFCD04065805
CAS Number: 62150-45-2
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR