Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-5-(trifluoromethyl)benzylamine ≥97%

Supplier: Alfa Aesar
Danger

H34423-06 H34423-03
AAH34423-06EA 420.5 USD InStock
AAH34423-06 AAH34423-03
3-Chloro-5-(trifluoromethyl)benzylamine ≥97%
3-Chloro-5-(trifluoromethyl)benzylamine


Formula: C₈H₇ClF₃N
MW: 209,60
Storage Temperature: Ambient
MDL Number: MFCD07786683
CAS Number: 400771-41-7
EINECS: 000-000-0
UN: 2735
ADR: 8,III
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty