N-(2-Bromophenyl)maleamic acid 97%

Warning

H34339-18 H34339-09
AAH34339-09EA 31.49 USD
AAH34339-09 AAH34339-18
N-(2-Bromophenyl)maleamic acid 97%
N-(2-Bromophenyl)maleamic acid
Formula: C₁₀H₈BrNO₃
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00157729
CAS Number: 59652-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR