3-Amino-4-fluorobenzonitrile 98%

Warning

H34336-03
AAH34336-03EA 108.44 USD
AAH34336-03
3-Amino-4-fluorobenzonitrile 98%
3-Amino-4-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787428
CAS Number: 859855-53-1
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR