2,6-Difluoro-4-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals

Warning

H34330-03 H34330-MD
AAH34330-03EA 180.86 USD
AAH34330-03 AAH34330-MD
2,6-Difluoro-4-methoxypyridine 97%
2,6-Difluoro-4-methoxypyridine
Formula: C₆H₅F₂NO
Storage Temperature: Ambient
MDL Number: MFCD13185564
CAS Number: 1184172-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR