3-Chloro-5-(pentafluorothio)aniline 97%

Warning

H34254-03 H34254-MD
AAH34254-03EA 275.28 USD
AAH34254-03 AAH34254-MD
3-Chloro-5-(pentafluorothio)aniline 97%
3-Chloro-5-(pentafluorothio)aniline
Formula: C₆H₅ClF₅NS
MW: 253.62
Storage Temperature: Ambient
MDL Number: MFCD16652453
CAS Number: 1240257-97-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR