3'-Fluoro-5'-(trifluoromethoxy)acetophenone 97%

Warning

H34207-03 H34207-MD
AAH34207-MDEA 54.46 USD
AAH34207-MD AAH34207-03
3'-Fluoro-5'-(trifluoromethoxy)acetophenone 97%
3'-Fluoro-5'-(trifluoromethoxy)acetophenone
Formula: C₉H₆F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18783230
CAS Number: 1352999-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR