3-Bromo-5-(pentafluorothio)benzoic acid 97%

Warning

H34194-03 H34194-MD
AAH34194-03EA 236.43 USD
AAH34194-03 AAH34194-MD
3-Bromo-5-(pentafluorothio)benzoic acid 97%
3-Bromo-5-(pentafluorothio)benzoic acid
Formula: C₇H₄BrF₅O₂S
MW: 247.16
Storage Temperature: Ambient
MDL Number: MFCD16652482
CAS Number: 1180675-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR