2-Fluoro-4-(pentafluorothio)benzoic acid 97%

Warning

H34180-06 H34180-03
AAH34180-06EA 418.38 USD
AAH34180-06 AAH34180-03
2-Fluoro-4-(pentafluorothio)benzoic acid 97%
2-Fluoro-4-(pentafluorothio)benzoic acid
Formula: C₇H₄F₆O₂S
MW: 266.16
Storage Temperature: Ambient
MDL Number: MFCD16652503
CAS Number: 1240257-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR