5-(2-Bromophenyl)isoxazole 95%

H34171-09 H34171-03
AAH34171-09EA 223.11 USD
AAH34171-09 AAH34171-03
5-(2-Bromophenyl)isoxazole 95%
5-(2-Bromophenyl)isoxazole
Formula: C₉H₆BrNO
MW: 224.06 g/mol
Boiling Pt: 95…97 °C/0.23 mm
Storage Temperature: Ambient
MDL Number: MFCD02183530
CAS Number: 387358-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR