Ethyl-5-fluorobenzo[b]thiophene-2-carboxylate 96%

H34167-03 H34167-MD H34167-06
AAH34167-03EA 102.75 USD
AAH34167-03 AAH34167-06 AAH34167-MD
Ethyl-5-fluorobenzo[b]thiophene-2-carboxylate 96%
Ethyl 5-fluorobenzo[b]thiophene-2-carboxylate
Formula: C₁₁H₉FO₂S
Melting Pt: 88…89 °C
Storage Temperature: Ambient
MDL Number: MFCD12923024
CAS Number: 13771-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR