1-Boc-4-iodopiperidine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl 4-iodopiperidine-1-carboxylate1-BOC-4-iodo-piperidine

H34090-06 H34090-03
AAH34090-03EA 110.99 USD
AAH34090-03 AAH34090-06
1-Boc-4-iodopiperidine 95%
1-Boc-4-iodopiperidine
Formula: C₁₀H₁₈INO₂
MW: 311.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115041
CAS Number: 301673-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR