4-(2-Methylphenoxy)aniline 96%

Warning

H34084-03
AAH34084-03EA 32.09 USD
AAH34084-03
4-(2-Methylphenoxy)aniline 96%
4-(2-Methylphenoxy)aniline
Formula: C₁₃H₁₃NO
Melting Pt: 61…62 °C
Storage Temperature: Ambient
EINECS: 260-349-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR