2',3',4',5',6'-Pentamethyl-2-phenylacetophenone 95%

Warning

Synonyms: 1-(2,3,4,5,6-Pentamethylphenyl)-2-phenylethan-1-one

H34080-09 H34080-03
AAH34080-03EA 33.74 USD
AAH34080-03 AAH34080-09
2',3',4',5',6'-Pentamethyl-2-phenylacetophenone 95%
2',3',4',5',6'-Pentamethyl-2-phenylacetophenone
Formula: C₁₉H₂₂O
MW: 266.38
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00052403
CAS Number: 147834-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR