Ethyl-2-(pyridin-3-yloxy)acetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (Pyridin-3-yloxy)acetic acid ethyl ester

H33981-03 H33981-09
AAH33981-03EA 32.59 USD
AAH33981-03 AAH33981-09
Ethyl-2-(pyridin-3-yloxy)acetate 98%
Ethyl-2-(pyridin-3-yloxy)acetate
Formula: C₉H₁₁NO₃
MW: 181.19
Density: 1.157
Storage Temperature: Ambient
MDL Number: MFCD00093962
CAS Number: 18342-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR