Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate 95%

Warning

H33977-MD H33977-03
AAH33977-MDEA 83.57 USD
AAH33977-MD AAH33977-03
Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate 95%
Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate
Formula: C₉H₉N₃O₂
MW: 191.19 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00847745
CAS Number: 115932-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR