2-Mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidin-4(3H)-one 96%

Warning

H33976-MD H33976-06 H33976-03
AAH33976-MDEA 23.45 USD
AAH33976-MD AAH33976-03 AAH33976-06
2-Mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidin-4(3H)-one 96%
2-Mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidin-4(3H)-one
Formula: C₁₇H₁₆N₂OS₂
MW: 328.46 g/mol
Melting Pt: 343…345 °C
Storage Temperature: Ambient
MDL Number: MFCD01896130
CAS Number: 299919-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR