2-Bromo-5-fluoro-3-methylpyridine 97%

Warning

H33953-03 H33953-06 H33953-MD
AAH33953-MDEA 49.49 USD
AAH33953-MD AAH33953-06 AAH33953-03
2-Bromo-5-fluoro-3-methylpyridine 97%
2-Bromo-5-fluoro-3-methylpyridine
Formula: C₆H₅BrFN
MW: 190.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835355
CAS Number: 38186-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR