2-Fluoro-4-(pentafluorothio)benzamide 97%

Warning

H33952-MD H33952-03
AAH33952-MDEA 67.49 USD
AAH33952-MD AAH33952-03
2-Fluoro-4-(pentafluorothio)benzamide 97%
2-Fluoro-4-(pentafluorothio)benzamide
Formula: C₇H₅F₆NOS
MW: 265.17
Storage Temperature: Ambient
MDL Number: MFCD16652504
CAS Number: 1240257-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR