1-Bromo-4-(difluoromethyl)benzene 95%

Warning

Synonyms: 4-Bromobenzal fluoride

H33941-03 H33941-06
AAH33941-03EA 102.96 USD
AAH33941-03 AAH33941-06
1-Bromo-4-(difluoromethyl)benzene 95%
1-Bromo-4-(difluoromethyl)benzene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 100 °C/53 mm
Storage Temperature: Ambient
MDL Number: MFCD06657964
CAS Number: 51776-71-7
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR