4-Methylsalicylaldehyde 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-4-methylbenzaldehyde

H33909-22 H33909-06 H33909-14
AAH33909-22EA 575.83 USD
AAH33909-22 AAH33909-14 AAH33909-06
4-Methylsalicylaldehyde 97+%
4-Methylsalicylaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 222…223 °C
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00799550
CAS Number: 698-27-1
EINECS: 211-813-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR