3-(3-Chlorophenylsulfonamido)benzoic acid 96%

Warning

H33885-03 H33885-MD
AAH33885-MDEA 33.63 USD
AAH33885-MD AAH33885-03
3-(3-Chlorophenylsulfonamido)benzoic acid 96%
3-(3-Chlorophenylsulfonamido)benzoic acid
Formula: C₁₃H₁₀ClNO₄S
MW: 311.75 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD06409487
CAS Number: 749884-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR