3-Chloro-5-(pentafluorosulfur)benzoic acid 97%

Warning

Synonyms: 3-Chloro-5-(pentafluorothio)benzoic acid

H33872-MD H33872-03
AAH33872-MDEA 34.34 USD
AAH33872-MD AAH33872-03
3-Chloro-5-(pentafluorosulfur)benzoic acid 97%
3-Chloro-5-(pentafluorosulfur)benzoic acid
Formula: C₇H₄ClF₅O₂S
MW: 282.615 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652415
CAS Number: 1211589-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR