2-Fluoro-4-(pentafluorothio)benzonitrile 97%

Warning

H33862-06 H33862-03
AAH33862-06EA 396.27 USD
AAH33862-06 AAH33862-03
2-Fluoro-4-(pentafluorothio)benzonitrile 97%
2-Fluoro-4-(pentafluorothio)benzonitrile
Formula: C₇H₃F₆NS
MW: 247.16
Storage Temperature: Ambient
MDL Number: MFCD16652501
CAS Number: 1240256-91-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR