4-Ethoxy-2-fluorobenzoyl chloride 97%

Danger

H33851-06 H33851-03
AAH33851-03EA 152.07 USD
AAH33851-03 AAH33851-06
4-Ethoxy-2-fluorobenzoyl chloride 97%
4-Ethoxy-2-fluorobenzoyl chloride
Formula: C₉H₈ClFO₂
MW: 202.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13196594
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR