Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 95%

Warning

Synonyms: 4,4,4,4',4',4'-Hexafluorovaline, ethyl ester

H33837-03 H33837-MD
AAH33837-MDEA 397.8 USD
AAH33837-MD AAH33837-03
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate 95%
Ethyl-2-amino-4,4,4-trifluoro-3-(trifluoromethyl)butyrate
Formula: C₇H₉F₆NO₂
MW: 253.15 g/mol
Boiling Pt: 70…72 °C/10 mm
Storage Temperature: Ambient
MDL Number: MFCD00040845
CAS Number: 78185-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR