3-Amino-2-bromo-5-fluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-5-fluoro-3-pyridylamine2-Bromo-5-fluoropyridin-3-amine

H33811-MD H33811-06 H33811-03
AAH33811-03EA 165.39 USD
AAH33811-03 AAH33811-06 AAH33811-MD
3-Amino-2-bromo-5-fluoropyridine 97%
3-Amino-2-bromo-5-fluoropyridine
Formula: C₅H₄BrFN₂
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD05662413
CAS Number: 884495-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR