1-Chloro-2,6-difluoro-3-nitrobenzene 97%

Danger

Synonyms: 3-Chloro-2,4-difluoronitrobenzene 2-Chloro-1,3-difluoro-4-nitrobenzene

H33806-14 H33806-03 H33806-06
AAH33806-06EA 126.54 USD
AAH33806-06 AAH33806-14 AAH33806-03
1-Chloro-2,6-difluoro-3-nitrobenzene 97%
1-Chloro-2,6-difluoro-3-nitrobenzene
Formula: C₆H₂ClF₂NO₂
MW: 193.54 g/mol
Melting Pt: 45…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00042201
CAS Number: 3847-58-3
EINECS: 411-980-8
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR