(2E)-3-(Dimethylamino)-1-(3-methoxyphenyl)prop-2-en-1-one 95%

Warning

H33797-14 H33797-06
AAH33797-06EA 72.9 USD
AAH33797-06 AAH33797-14
(2E)-3-(Dimethylamino)-1-(3-methoxyphenyl)prop-2-en-1-one 95%
(2E)-3-(Dimethylamino)-1-(3-methoxyphenyl)prop-2-en-1-one
Formula: C₁₂H₁₅NO₂
Melting Pt: 46…48 °C
Storage Temperature: Ambient
MDL Number: MFCD11501639
CAS Number: 62041-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR