4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine 96%

Supplier: Thermo Scientific Chemicals
Warning

H33770-03 H33770-06 H33770-MD
AAH33770-03EA 124.99 USD
AAH33770-03 AAH33770-06 AAH33770-MD
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine 96%
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine
Formula: C₈H₇ClN₂S
Melting Pt: 109…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00464832
CAS Number: 108831-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR