4-Ethoxy-2-fluorobenzonitrile 97%

Warning

H33730-06 H33730-03
AAH33730-06EA 396.27 USD
AAH33730-06 AAH33730-03
4-Ethoxy-2-fluorobenzonitrile 97%
4-Ethoxy-2-fluorobenzonitrile
Formula: C₉H₈FNO
Storage Temperature: Ambient
MDL Number: MFCD13196591
CAS Number: 186191-34-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR