Methyl-2-[(chloroacetyl)amino]-4-phenylthiophene-3-carboxylate 96%

Warning

H33721-03 H33721-06
AAH33721-03EA 29.5 USD
AAH33721-03 AAH33721-06
Methyl-2-[(chloroacetyl)amino]-4-phenylthiophene-3-carboxylate 96%
Methyl-2-[(chloroacetyl)amino]-4-phenylthiophene-3-carboxylate
Formula: C₁₄H₁₂ClNO₃S
Melting Pt: 107…110 °C
Storage Temperature: Ambient
MDL Number: MFCD01917476
CAS Number: 356568-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR