2-Fluoro-4-(trifluoromethoxy)phenylacetonitrile 97%

Warning

H33686-MD H33686-03
AAH33686-03EA 269.73 USD
AAH33686-03 AAH33686-MD
2-Fluoro-4-(trifluoromethoxy)phenylacetonitrile 97%
2-Fluoro-4-(trifluoromethoxy)phenylacetonitrile
Formula: C₉H₅F₄NO
MW: 219.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16652443
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR