2-Nitropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H33657-06
H33657-03
AAH33657-06EA
163.99
USD
AAH33657-06
AAH33657-03
2-Nitropyridine 97%
2-Nitropyridine
Formula:
C₅H₄N₂O₂ MW: 124.10 g/mol Melting Pt: 69…73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00160414 CAS Number: 15009-91-3 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...