2-Fluoro-4-(trifluoromethoxy)benzylamine 97%

Danger

H33649-MD H33649-03
AAH33649-03EA 158.73 USD
AAH33649-03 AAH33649-MD
2-Fluoro-4-(trifluoromethoxy)benzylamine 97%
2-Fluoro-4-(trifluoromethoxy)benzylamine
Formula: C₈H₇F₄NO
Storage Temperature: Ambient
MDL Number: MFCD16652498
CAS Number: 1240257-11-7
UN: 3267
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR