1-Chloro-4-(3,4-dimethylphenyl)phthalazine 96%

Warning

H33635-MD H33635-06 H33635-03
AAH33635-MDEA 21.35 USD
AAH33635-MD AAH33635-03 AAH33635-06
1-Chloro-4-(3,4-dimethylphenyl)phthalazine 96%
1-Chloro-4-(3,4-dimethylphenyl)phthalazine
Formula: C₁₆H₁₃ClN₂
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD03001385
CAS Number: 129842-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR