2,3,5-Tri-O-benzyl-D-ribofuranose 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2R,3R,4R)-2,3,5-Tris(benzyloxy)-4-hydroxypentanal

H33605-03 H33605-06
AAH33605-06EA 291.99 USD
AAH33605-06 AAH33605-03
2,3,5-Tri-O-benzyl-D-ribofuranose 98%
2,3,5-Tri-O-benzyl-D-ribofuranose
Formula: C₂₆H₂₈O₅
MW: 420.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425640
CAS Number: 54623-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR