4-(2-Methylphenoxy)benzenesulfonyl chloride 96%

Danger

H33556-06 H33556-MD H33556-03
AAH33556-06EA 327.2 USD
AAH33556-06 AAH33556-MD AAH33556-03
4-(2-Methylphenoxy)benzenesulfonyl chloride 96%
4-(2-Methylphenoxy)benzenesulfonyl chloride
Formula: C₁₃H₁₁ClO₃S
MW: 282.74 g/mol
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD01631900
CAS Number: 610277-83-3
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR