4-Fluorobenzoic anhydride 97%

Supplier: Thermo Scientific Chemicals

Warning

Synonyms: 4-fluorobenzene-1-carboxylic anhydride

H33527-03 H33527-06
AAH33527-03EA 89.85 USD
AAH33527-03 AAH33527-06
4-Fluorobenzoic anhydride 97%
4-Fluorobenzoic anhydride
Formula: C₁₄H₈F₂O₃
MW: 262.21 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00425938
CAS Number: 25569-77-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR