2-Fluoro-5-(pentafluorothio)benzaldehyde 97%

Warning

H33481-MD H33481-03
AAH33481-03EA 236.43 USD
AAH33481-03 AAH33481-MD
2-Fluoro-5-(pentafluorothio)benzaldehyde 97%
2-Fluoro-5-(pentafluorothio)benzaldehyde
Formula: C₇H₄F₆OS
MW: 250.16
Storage Temperature: Ambient
MDL Number: MFCD16652493
CAS Number: 1240257-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR