3-(2-Furyl)-1H-pyrazole 97%

Warning

H33472-03 H33472-MD H33472-06
AAH33472-03EA 62.72 USD
AAH33472-03 AAH33472-06 AAH33472-MD
3-(2-Furyl)-1H-pyrazole 97%
3-(2-Furyl)-1H-pyrazole
Formula: C₇H₆N₂O
MW: 134.14 g/mol
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00973923
CAS Number: 32332-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR