3-Fluoro-5-(pentafluorothio)benzaldehyde 97%

Warning

H33427-MD H33427-03
AAH33427-MDEA 24.63 USD
AAH33427-MD AAH33427-03
3-Fluoro-5-(pentafluorothio)benzaldehyde 97%
3-Fluoro-5-(pentafluorothio)benzaldehyde
Formula: C₇H₄F₆OS
MW: 250.16
Storage Temperature: Ambient
MDL Number: MFCD16652417
CAS Number: 1240257-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR