2,5-Bis(trifluoromethyl)thiobenzamide 97%
|
Warning
|
H33419-03
H33419-MD
AAH33419-03EA
175.92
USD
AAH33419-03
AAH33419-MD
2,5-Bis(trifluoromethyl)thiobenzamide 97%
2,5-Bis(trifluoromethyl)thiobenzamide
Formula:
C₉H₅F₆NS Melting Pt: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD09757524 CAS Number: 1186194-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...